Assay Details
| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110639.1 | 1 - 2 | 234 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein unc-80 homolog | LOC103693241 | -44.7kb | Sense | Chr. 9 - 72663790 - 72665685 on NCBI Build Rnor_6.0 | |
| Downstream | tensin 1 | Tns1 | Tns | 494.4kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC287004
Genomic Map