Assay Details
| Gene Symbol: Terc | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001567.1 | LIT2120 telomerase RNA |
1 | 245 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | actin-related protein T3 | Actrt3 |
Arpm1, RGD1561457 |
-14.2kb | Sense | Chr. 2 - 116416561 - 116418521 on NCBI Build Rnor_6.0 |
| Downstream | EGF-like and EMI domain containing 1 | Egfem1 | RGD1307225 | 93.0kb | Sense | Chr. 2 - 116526140 - 117152634 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Terc
Genomic Map