Assay Details
| Gene Symbol: LOC287004 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110637.1 | 1 | 2350 | 108 | |||
| GenBank mRNA | DQ080211.1 | - | LIT3459 PINC |
1 | 2350 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transition protein 1 | Tnp1 | -392.3kb | Sense | Chr. 9 - 80294837 - 80295446 on NCBI Build Rnor_6.0 | |
| Downstream | tensin 1 | Tns1 | Tns | 501.4kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC287004
Genomic Map