Assay Details
| Gene Symbol: Tigd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126268.2 | NP_001119740.1 | 1 | 1749 | 80 | |
| GenBank mRNA | BC161993.1 | - | 1 | 1749 | 9033247 | 80 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tigd5
Genomic Map