Assay Details
| Gene Symbol: Zfp955a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126281.1 | NP_001119753.1 | 2 - 3 | 223 | 89 | |
| GenBank mRNA | BC160923.1 | - | 2 - 3 | 223 | 8362600 | 89 |
| BC168190.1 | - | 2 - 3 | 191 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp955a
Genomic Map