Assay Details
| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135874.1 | NP_001129346.1 | 2 - 3 | 487 | 70 | |
| XM_006240406.2 | XP_006240468.1 | 3 - 4 | 984 | 70 | ||
| XM_006240407.2 | XP_006240469.1 | 3 - 4 | 722 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ston2
Genomic Map