Assay Details
| Gene Symbol: Ube2q2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135997.2 | NP_001129469.1 | 1 | 717 | 80 | |
| GenBank mRNA | FQ225248.1 | - | 1 | 164 | 80 | |
| FQ225615.1 | - | 1 | 164 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2q2l
Genomic Map