Assay Details
| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109607.1 | NP_001103077.1 | 1 | 45 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109607.1 transcripts.
Set Membership: |
|

Hint1
Genomic Map