Assay Details
| Gene Symbol: Defb16-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002707.1 | 1 - 2 | 51 | 102 | ||
| GenBank mRNA | DQ012089.1 | - | 1 - 2 | 51 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | defensin beta 17 | Defb17 | Sense | Chr. 9 - 24506246 - 24509761 on NCBI Build Rnor_6.0 | ||
| Downstream | transcription factor AP-2 delta | Tfap2d | Tcfap2d | 795.2kb | Anti-sense | Chr. 9 - 25304876 - 25365151 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Defb16-ps
Genomic Map