Assay Details
| Gene Symbol: RT1-T24-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002151.1 | 1 | 1951 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RT1 class I histocompatibility antigen, AA alpha chain-like |
LOC100909797 | Anti-sense | Chr. 20 - 3242236 - 3253995 on NCBI Build Rnor_6.0 | ||
| Downstream |
RT1 class I, locus T24, gene 1 |
RT1-T24-1 | H2-T24 | 22.1kb | Anti-sense | Chr. 20 - 3269062 - 3289057 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RT1-T24-2
Genomic Map