Assay Details
| Gene Symbol: RGD1562963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145021.1 | NP_001138493.1 | 2 - 3 | 324 | 81 | |
| XM_006253780.2 | XP_006253842.1 | 2 - 3 | 926 | 81 | ||
| GenBank mRNA | BC098763.1 | - | 2 - 3 | 324 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1562963
Genomic Map