Assay Details
| Gene Symbol: Tbcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108200.2 | NP_001101670.1 | 1 | 846 | 111 | |
| GenBank mRNA | BC166467.1 | - | 1 | 846 | 9035211 | 111 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbcc
Genomic Map