Assay Details
| Gene Symbol: LOC257642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147136.1 | NP_671477.1 | 752 | 105 | ||
| GenBank mRNA | AY539885.1 | - | 347 | 105 | ||
| U77931.1 | - | 752 | 105 | |||
| Gene Symbol: LOC103693757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770183.1 | XP_008768405.1 | 4 | 2272 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC257642
Genomic Map