Assay Details
| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750021.3 | XP_003750069.1 | 2 - 3 | 225 | 86 | |
| XM_008764092.1 | XP_008762314.1 | 245 | 86 | |||
More Information
Set Membership: |
|

Cfap57
Genomic Map