Assay Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165936.1 | NP_001159408.1 | 4 - 5 | 675 | 58 | |
| NM_212532.2 | NP_997697.1 | 4 - 5 | 722 | 58 | ||
| XM_008772714.1 | XP_008770936.1 | 5 - 6 | 821 | 58 | ||
| XM_008772715.1 | XP_008770937.1 | 4 - 5 | 984 | 58 | ||
| GenBank mRNA | BC086443.1 | - | 4 - 5 | 623 | 7303579 | 58 |
| BC097930.1 | - | 4 - 5 | 712 | 7388716 | 58 | |
| CO385374.1 | - | 4 - 5 | 722 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddah2
Genomic Map