Assay Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017196.3 | NP_058892.1 | 5 - 6 | 451 | 82 | |
| XM_006256061.2 | XP_006256123.1 | 4 - 5 | 825 | 82 | ||
| XM_006256062.2 | XP_006256124.1 | 5 - 6 | 802 | 82 | ||
| XM_006256063.2 | XP_006256125.1 | 5 - 6 | 765 | 82 | ||
| XM_006256065.2 | XP_006256127.1 | 7 - 8 | 601 | 82 | ||
| GenBank mRNA | AB000818.1 | - | 5 - 6 | 449 | 82 | |
| AF299328.1 | - | 2 - 3 | 191 | 82 | ||
| BC081822.1 | - | 3 - 4 | 1298 | 82 | ||
| D82069.1 | - | 7 - 8 | 606 | 82 | ||
| FM053942.1 | - | 5 - 6 | 438 | 82 | ||
| FQ231499.1 | - | 5 - 6 | 449 | 82 | ||
| U17919.1 | - | 5 - 6 | 423 | 82 | ||
More Information
Set Membership: |
|

Aif1
Genomic Map