Assay Details
| Gene Symbol: Lingo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100722.1 | NP_001094192.1 | 2 | 1855 | 101 | |
| XM_006243120.2 | XP_006243182.1 | 2 | 2978 | 101 | ||
| XM_006243121.2 | XP_006243183.1 | 2 | 2007 | 101 | ||
| XM_008766270.1 | XP_008764492.1 | 2 | 2005 | 101 | ||
| XM_008766271.1 | XP_008764493.1 | 3 | 2099 | 101 | ||
| GenBank mRNA | BC092632.1 | - | 1 | 1314 | 101 | |
More Information
Set Membership: |
|

Lingo1
Genomic Map