Assay Details
| Gene Symbol: Ttll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169136.1 | NP_001162607.1 | 1 - 2 | 77 | 92 | |
| XM_006227919.2 | XP_006227981.1 | 2 - 3 | 3374 | 92 | ||
| XM_006227921.2 | XP_006227983.1 | 2 - 3 | 3566 | 92 | ||
| XM_008758751.1 | XP_008756973.1 | 2 - 3 | 3377 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll2
Genomic Map