Assay Details
Gene Symbol: Olr1587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000913.1 | NP_001000913.1 | 1 | 553 | 67 | |
XM_008769791.1 | XP_008768013.1 | 2 | 3523 | 67 | ||
XM_008769792.1 | XP_008768014.1 | 3 | 3265 | 67 | ||
XM_008769793.1 | XP_008768015.1 | 3 | 3266 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
