Assay Details
| Gene Symbol: Vom1r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009517.1 | NP_001009517.1 | 1 | 539 | 101 | |
| GenBank mRNA | AY510342.1 | - | 1 | 539 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vom1r3
Genomic Map