Assay Details
| Gene Symbol: Ptges3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014272.1 | NP_001014294.1 | 1 - 2 | 27 | 84 | |
| GenBank mRNA | BC087125.1 | - | 1 - 2 | 27 | 7110309 | 84 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC087125.1,NM_001014272.1 transcripts.
Set Membership: |
|

Ptges3l1
Genomic Map