Assay Details
| Gene Symbol: Pdhx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044242.1 | NP_001037707.1 | 8 - 9 | 1026 | 65 | |
| GenBank mRNA | AY310145.1 | - | 8 - 9 | 1026 | 65 | |
| BC091292.1 | - | 5 - 6 | 663 | 65 | ||
| FQ232219.1 | - | 1053 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pdhx
Genomic Map