Assay Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100885.1 | NP_001094355.1 | 1 - 2 | 194 | 86 | |
| XM_006235401.1 | XP_006235463.1 | 2 - 3 | 310 | 86 | ||
| XM_006235402.2 | XP_006235464.1 | 2 - 3 | 324 | 86 | ||
| GenBank mRNA | BC158548.1 | - | 2 - 3 | 273 | 9036351 | 86 |
| FQ215756.1 | - | 2 - 3 | 247 | 86 | ||
| FQ219977.1 | - | 2 - 3 | 251 | 86 | ||
| FQ220245.1 | - | 2 - 3 | 251 | 86 | ||
| FQ220303.1 | - | 2 - 3 | 251 | 86 | ||
| FQ220687.1 | - | 2 - 3 | 253 | 86 | ||
| FQ227515.1 | - | 2 - 3 | 254 | 86 | ||
| FQ229420.1 | - | 2 - 3 | 251 | 86 | ||
| FQ229727.1 | - | 2 - 3 | 254 | 86 | ||
| FQ229762.1 | - | 2 - 3 | 253 | 86 | ||
| FQ229855.1 | - | 2 - 3 | 287 | 86 | ||
| FQ230407.1 | - | 2 - 3 | 267 | 86 | ||
| FQ231493.1 | - | 2 - 3 | 267 | 86 | ||
| FQ231820.1 | - | 2 - 3 | 286 | 86 | ||
| FQ232091.1 | - | 2 - 3 | 247 | 86 | ||
| S77900.1 | - | 1 - 2 | 194 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Myl9
Genomic Map