Assay Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127375.1 | NP_001120847.1 | 1 - 2 | 160 | 106 | |
| GenBank mRNA | BC166476.1 | - | 1 - 2 | 160 | 9101314 | 106 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166476.1, NM_001127375.1 transcripts.
Set Membership: |
|

Zbtb7c
Genomic Map