Assay Details
| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165897.1 | NP_001159369.1 | 1 - 2 | 103 | 91 | |
| GenBank mRNA | CK478392.1 | - | 1 - 2 | 91 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001165897.1 transcripts.
Set Membership: |
|

Tor1aip2
Genomic Map