Assay Details
| Gene Symbol: Tmem11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166165.1 | NP_001159637.1 | 2 - 3 | 591 | 81 | |
| GenBank mRNA | DY316208.1 | - | 2 - 3 | 334 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001166165.1 transcripts.
This assay is in the 5'UTR region of the NM_001166165.1 transcripts.
Set Membership: |
|

Tmem11
Genomic Map