Assay Details
| Gene Symbol: Id3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013058.2 | NP_037190.1 | 1 - 2 | 337 | 80 | |
| GenBank mRNA | BC064658.1 | - | 1 - 2 | 337 | 5623701 | 80 |
| D10864.1 | - | 1 - 2 | 314 | 80 | ||
| FQ212997.1 | - | 1 - 2 | 359 | 80 | ||
| FQ216267.1 | - | 1 - 2 | 359 | 80 | ||
| FQ218468.1 | - | 1 - 2 | 357 | 80 | ||
| FQ218958.1 | - | 1 - 2 | 350 | 80 | ||
| FQ229332.1 | - | 1 - 2 | 359 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Id3
Genomic Map