Assay Details
| Gene Symbol: Kcnj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017296.1 | NP_058992.1 | 1 | 1145 | 107 | |
| XM_006247567.2 | XP_006247629.1 | 2 | 4478 | 107 | ||
| GenBank mRNA | AF021137.1 | - | 1 | 1145 | 107 | |
| L48490.1 | - | 1 | 1145 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kcnj2
Genomic Map