Assay Details
| Gene Symbol: Nmur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022275.3 | NP_071611.3 | 2 - 3 | 915 | 95 | |
| GenBank mRNA | AB041229.1 | - | 2 - 3 | 915 | 95 | |
| AF242875.1 | - | 2 - 3 | 789 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nmur2
Genomic Map