Assay Details
Gene Symbol: Kcnj10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031602.1 | NP_113790.1 | 1 - 2 | 118 | 109 | |
GenBank mRNA | U27558.1 | - | 1 - 2 | 118 | 109 | |
X86818.1 | - | 1 - 2 | 116 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_031602.1, U27558.1, X86818.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
