Assay Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053843.2 | NP_446295.2 | 1 - 2 | 197 | 78 | |
GenBank mRNA | L08446.1 | - | 1 - 2 | 197 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L08446.1, NM_053843.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
