Assay Details
| Gene Symbol: Nat8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133558.1 | NP_598242.1 | 71 | 67 | ||
| GenBank mRNA | AF163318.1 | - | 71 | 67 | ||
| BC081733.1 | - | 121 | 7109438 | 67 | ||
| Gene Symbol: LOC103690139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763104.1 | XP_008761326.1 | 1 - 2 | 443 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF163318.1, BC081733.1, NM_133558.1, XM_008763104.1 transcripts.
Set Membership: |
|

Nat8b
Genomic Map