Assay Details
| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139108.2 | NP_620808.1 | 69 | 64 | ||
| GenBank mRNA | AF290194.1 | - | 74 | 64 | ||
| Gene Symbol: LOC100910472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241916.2 | XP_006241978.1 | 1 - 2 | 398 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF290194.1, NM_139108.2, XM_006241916.2 transcripts.
Set Membership: |
|

Commd5
Genomic Map