Assay Details
| Gene Symbol: Npc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173118.2 | NP_775141.2 | 2 - 3 | 294 | 64 | |
| GenBank mRNA | AJ515237.1 | - | 2 - 3 | 229 | 64 | |
| BC058132.1 | - | 2 - 3 | 257 | 6920130 | 64 | |
| FQ214040.1 | - | 2 - 3 | 215 | 64 | ||
| FQ216545.1 | - | 2 - 3 | 219 | 64 | ||
| FQ219301.1 | - | 2 - 3 | 236 | 64 | ||
| FQ219863.1 | - | 2 - 3 | 227 | 64 | ||
| FQ220660.1 | - | 2 - 3 | 215 | 64 | ||
| FQ220919.1 | - | 2 - 3 | 221 | 64 | ||
| FQ228456.1 | - | 2 - 3 | 219 | 64 | ||
| FQ229422.1 | - | 2 - 3 | 274 | 64 | ||
| FQ229782.1 | - | 2 - 3 | 215 | 64 | ||
| FQ229783.1 | - | 2 - 3 | 217 | 64 | ||
| FQ230107.1 | - | 2 - 3 | 221 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npc2
Genomic Map