Assay Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173142.1 | NP_775165.1 | 1 - 2 | 192 | 85 | |
| XM_006245160.1 | XP_006245222.1 | 1 - 2 | 195 | 85 | ||
| GenBank mRNA | D86220.1 | - | 1 - 2 | 192 | 85 | |
| FQ213827.1 | - | 1 - 2 | 188 | 85 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the D86220.1,NM_173142.1,XM_006245160.1 transcripts.
Set Membership: |
|

Stk16
Genomic Map