Assay Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173142.1 | NP_775165.1 | 4 - 5 | 724 | 70 | |
| XM_006245160.1 | XP_006245222.1 | 4 - 5 | 727 | 70 | ||
| GenBank mRNA | D86220.1 | - | 4 - 5 | 724 | 70 | |
| FQ213827.1 | - | 4 - 5 | 720 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk16
Genomic Map