Assay Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173142.1 | NP_775165.1 | 6 - 7 | 938 | 79 | |
| GenBank mRNA | D86220.1 | - | 6 - 7 | 938 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk16
Genomic Map