Assay Details
| Gene Symbol: Rup2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034950.2 | NP_001030122.2 | 2 - 3 | 235 | 96 | |
| GenBank mRNA | AF198441.1 | - | 2 - 3 | 207 | 96 | |
| AY327506.1 | - | 2 - 3 | 235 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rup2
Genomic Map