Assay Details
| Gene Symbol: Lilra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076793.1 | NP_001070261.1 | 4 - 5 | 665 | 95 | |
| XM_006228225.2 | XP_006228287.1 | 3 - 4 | 886 | 95 | ||
| XM_008758988.1 | XP_008757210.1 | 4 - 5 | 688 | 95 | ||
| XM_008758989.1 | XP_008757211.1 | 3 - 4 | 652 | 95 | ||
| GenBank mRNA | AY998481.1 | - | 4 - 5 | 665 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lilra5
Genomic Map