Assay Details
| Gene Symbol: Rpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077592.1 | NP_001071060.1 | 2 - 3 | 56 | 92 | |
| GenBank mRNA | BC101933.1 | - | 2 - 3 | 56 | 7371608 | 92 |
| FQ210073.1 | - | 2 - 3 | 63 | 92 | ||
| FQ217659.1 | - | 2 - 3 | 58 | 92 | ||
| FQ222790.1 | - | 2 - 3 | 63 | 92 | ||
| FQ223953.1 | - | 2 - 3 | 63 | 92 | ||
| FQ224522.1 | - | 2 - 3 | 92 | |||
| FQ228832.1 | - | 2 - 3 | 74 | 92 | ||
| X57007.1 | - | 2 - 3 | 91 | |||
| Gene Symbol: LOC685963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001065954.5 | XP_001065954.1 | 1 | 78 | 92 | |
| Gene Symbol: LOC690468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001074532.4 | XP_001074532.1 | 1 | 60 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC101933.1,NM_001077592.1,X57007.1,XM_001065954.5,XM_001074532.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl38
Genomic Map