Assay Details
| Gene Symbol: Rpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077592.1 | NP_001071060.1 | 3 - 4 | 114 | 99 | |
| XM_008768393.1 | XP_008766615.1 | 3 - 4 | 113 | 99 | ||
| GenBank mRNA | BC101933.1 | - | 3 - 4 | 114 | 7371608 | 99 |
| FQ217659.1 | - | 3 - 4 | 116 | 99 | ||
| FQ223953.1 | - | 3 - 4 | 121 | 99 | ||
| FQ224522.1 | - | 3 - 4 | 121 | 99 | ||
| FQ228832.1 | - | 3 - 4 | 132 | 99 | ||
| X57007.1 | - | 3 - 4 | 119 | 99 | ||
| Gene Symbol: LOC685963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001065954.5 | XP_001065954.1 | 1 | 136 | 99 | |
| Gene Symbol: LOC690468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001074532.4 | XP_001074532.1 | 1 | 118 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl38
Genomic Map