Assay Details
| Gene Symbol: Dusp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079893.2 | NP_001073362.1 | 2 | 636 | 102 | |
| NM_001270835.1 | NP_001257764.1 | 2 | 623 | 102 | ||
| NM_001270836.1 | NP_001257765.1 | 2 | 627 | 102 | ||
| GenBank mRNA | BC158555.1 | - | 2 | 605 | 7311099 | 102 |
| EF122004.1 | - | 1 | 449 | 102 | ||
| EF122005.1 | - | 1 | 449 | 102 | ||
| FQ213995.1 | - | 2 | 623 | 102 | ||
| Gene Symbol: Dusp14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749277.3 | XP_003749325.1 | 1 | 662 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dusp14
Genomic Map