Assay Details
| Gene Symbol: Kcnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033376.1 | NP_203694.1 | 1 - 2 | 383 | 72 | |
| GenBank mRNA | AB048823.1 | - | 2 - 3 | 236 | 72 | |
| AB048824.1 | - | 2 - 3 | 225 | 72 | ||
| AF031384.1 | - | 1 - 2 | 383 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB048824.1 transcripts.
Set Membership: |
|

Kcnk3
Genomic Map