Assay Details
| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199208.1 | NP_954678.1 | 961 | 111 | ||
| GenBank mRNA | BC062000.1 | - | 928 | 5599919 | 111 | |
| BC079153.1 | - | 3 - 4 | 785 | 7128686 | 111 | |
| U33500.1 | - | 961 | 111 | |||
| Gene Symbol: LOC100365958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241512.2 | XP_006241574.1 | 3 - 4 | 939 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rdh16
Genomic Map