Assay Details
| Gene Symbol: Gstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024304.1 | NP_001019475.1 | 1 | 50 | 69 | |
| GenBank mRNA | BC091199.1 | - | 1 | 50 | 7379741 | 69 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC091199.1,NM_001024304.1 transcripts.
This assay is in the 5'UTR region of the BC091199.1,NM_001024304.1 transcripts.
Set Membership: |
|

Gstm4
Genomic Map