Assay Details
| Gene Symbol: Klk1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012677.1 | NP_036809.1 | 1 | 53 | 79 | |
| GenBank mRNA | M11565.1 | - | 1 | 53 | 79 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the M11565.1,NM_012677.1 transcripts.
Set Membership: |
|

Klk1c2
Genomic Map