Assay Details
Gene Symbol: Pcdha13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053934.2 | NP_446386.2 | 1 | 2053 | 110 | |
GenBank mRNA | AB045586.1 | - | 1 | 1331 | 110 | |
AB113396.1 | - | 1 | 2053 | 110 | ||
AY573975.1 | - | 1 | 2053 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
