Assay Details
| Gene Symbol: Fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013146.1 | NP_001013164.1 | 3 | 352 | 93 | |
| GenBank mRNA | AY622611.1 | - | 4 | 604 | 93 | |
| U18982.1 | - | 3 | 352 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fosl2
Genomic Map