Assay Details
| Gene Symbol: Mir196a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031913.1 | 1 | 57 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeo box C10 | Hoxc10 | -1.4kb | Sense | Chr. 7 - 144577512 - 144582802 on NCBI Build Rnor_6.0 | |
| Downstream | homeobox C9 | Hoxc9 | Hoxc8 | 8.1kb | Sense | Chr. 7 - 144592470 - 144598807 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir196a
Genomic Map