Assay Details
| Gene Symbol: Mir433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031954.1 | 1 | 45 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 431 | Mir431 | rno-mir-431 | -796.0 | Sense | Chr. 6 - 133711425 - 133711538 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 127 | Mir127 | rno-mir-127 | 1.0kb | Sense | Chr. 6 - 133713430 - 133713526 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir433
Genomic Map